5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Supplier: Apollo Scientific
Warning
PC6318-1G PC6318-250MG
APOSPC6318-1GEA 77 EUR
APOSPC6318-1G APOSPC6318-250MG
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Formula: C₆H₄ClF₃N₂O₂
MW: 228.56 g/mol
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00215443
CAS Number: 128455-63-0