5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
Supplier: Apollo Scientific
PC6325-1G PC6325-500MG PC6325-5G
APOSPC6325-1GEA 533 EUR
APOSPC6325-1G APOSPC6325-500MG APOSPC6325-5G
5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
5-(3-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formula: C₁₂H₈ClF₃N₂O₂
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00243688
CAS Number: 282523-36-8