[5-[(4-Chlorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%
Supplier: Apollo Scientific
PC6336-1G PC6336-500MG PC6336-5G
APOSPC6336-1GEA 533 EUR
APOSPC6336-1G APOSPC6336-500MG APOSPC6336-5G
[5-[(4-Chlorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 97%
[5-[(4-Chlorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Formula: C₁₂H₁₀ClF₃N₂OS
MW: 322.74 g/mol
Melting Pt: 96…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00172635
CAS Number: 318239-52-0