Silver fluoride 98%
Supplier: Apollo Scientific
Danger
Synonyms: Silver monofluoride, Silver(I) fluoride, Argentous fluoride
PC6370-5G PC6370-25G PC6370-1G
APOSPC6370-5GEA 79.9 EUR
APOSPC6370-5G APOSPC6370-25G APOSPC6370-1G
Silver fluoride 98%
Silver fluoride
Soft (mild) fluorinating agent for selective fluorination. Alkyl fluorides may be prepared by halex reactions. AgF & I2 or Br2 will trans-fluoro halogenate alkenes.
Formula: AgF
MW: 126.87 g/mol
Boiling Pt: 1159 °C
Melting Pt: >300 °C
Density: 5.852 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003410
CAS Number: 7775-41-9
UN: 3260
ADR: 8,II