2-Fluoro-DL-tyrosine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-3-(2-fluoro-4-hydroxyphenyl)propanoic acid, H-DL-Tyr(2-F)-OH
PC6380-1G PC6380-250MG
APOSPC6380-1GEA 835 EUR
APOSPC6380-1G APOSPC6380-250MG
2-Fluoro-DL-tyrosine 97%
2-Fluoro-DL-tyrosine
Formula: C₉H₁₀FNO₃
MW: 199.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03005295
CAS Number: 7656-31-7