3-Fluoro-4-(trifluoromethoxy)benzonitrile
Supplier: Apollo Scientific
Warning
PC6423-1G PC6423-25G PC6423-5G
APOSPC6423-1GEA 42.2 EUR
APOSPC6423-1G APOSPC6423-25G APOSPC6423-5G
3-Fluoro-4-(trifluoromethoxy)benzonitrile
3-Fluoro-4-(trifluoromethoxy)benzonitrile
Formula: C₈H₃F₄NO
Boiling Pt: 60 °C/ 5mmHg
Storage Temperature: Ambient
MDL Number: MFCD06660203
CAS Number: 886498-94-8
UN: 3276
ADR: 6.1,III