Methyl-3-cyano-4-fluorobenzoate 98+%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-cyano-4-fluorobenzoate
PC6425-1G PC6425-25G PC6425-5G
APOSPC6425-1GEA 21.9 EUR
APOSPC6425-1G APOSPC6425-25G APOSPC6425-5G
Methyl-3-cyano-4-fluorobenzoate 98+%
Methyl-3-cyano-4-fluorobenzoate
Formula: C₉H₆FNO₂
MW: 179.1500 g/mol
Melting Pt: 88…91 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06203915
CAS Number: 676602-31-6
UN: 3439
ADR: 6.1 III