6-Fluoro-4-hydroxycoumarin
Supplier: Apollo Scientific
Warning
Synonyms: 6-Fluoro-4-hydroxy-2H-chromen-2-one
PC6507-1G PC6507-250MG
APOSPC6507-1GEA 43.5 EUR
APOSPC6507-1G APOSPC6507-250MG
6-Fluoro-4-hydroxycoumarin
6-Fluoro-4-hydroxycoumarin
Formula: C₉H₅FO₃
MW: 180.13 g/mol
Melting Pt: 248…251 °C
Storage Temperature: Ambient
MDL Number: MFCD03094006
CAS Number: 1994-13-4