5-(Trifluoromethoxy)salicylaldehyde 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
PC6513-1G PC6513-25G PC6513-5G
APOSPC6513-1GEA 21.9 EUR
APOSPC6513-1G APOSPC6513-25G APOSPC6513-5G
5-(Trifluoromethoxy)salicylaldehyde 99%
5-(Trifluoromethoxy)salicylaldehyde
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 82 °C/60mmHg
Melting Pt: 31…33 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075249
CAS Number: 93249-62-8