2,6-Dichloro-4-(trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
Warning
PC6533-10G PC6533-5G
APOSPC6533-10GEA 31.9 EUR
APOSPC6533-10G APOSPC6533-5G
2,6-Dichloro-4-(trifluoromethoxy)aniline 98%
2,6-Dichloro-4-(trifluoromethoxy)aniline
Formula: C₇H₄Cl₂F₃NO
Melting Pt: 30…32 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00190128
CAS Number: 99479-66-0