4-Nitro-3-(trifluoromethyl)benzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 4-Nitro-3-(trifluoromethyl)benzene-1-sulphonyl chloride
PC6567-1G PC6567-5G
APOSPC6567-1GEA 102 EUR
APOSPC6567-1G APOSPC6567-5G
4-Nitro-3-(trifluoromethyl)benzenesulphonyl chloride
4-Nitro-3-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃ClF₃NO₄S
MW: 289.62 g/mol
Melting Pt: 61…67 °C
Storage Temperature: Ambient
MDL Number: MFCD03094387
CAS Number: 39234-83-8
UN: 3261
ADR: 8 II