3-Fluoro-4-methylbenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Fluoro-4-methylbenzene-1-sulphonyl chloride
PC6569-5G PC6569-25G
APOSPC6569-5GEA 79.9 EUR
APOSPC6569-5G APOSPC6569-25G
3-Fluoro-4-methylbenzenesulphonyl chloride 97%
3-Fluoro-4-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094389
CAS Number: 90260-13-2
UN: 3261
ADR: 8 II