4-Fluoro-2-methylbenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 4-Fluoro-2-methylbenzene-1-sulphonyl chloride
PC6571-25G PC6571-5G
APOSPC6571-25GEA 422 EUR
APOSPC6571-25G APOSPC6571-5G
4-Fluoro-2-methylbenzenesulphonyl chloride
4-Fluoro-2-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
Boiling Pt: 245…246 °C
Density: 1.433 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094390
CAS Number: 7079-48-3
UN: 3265
ADR: 8,II