4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Bromo-2-(trifluoromethyl)benzene-1-sulphonyl chloride
PC6576-100G PC6576-25G PC6576-5G
APOSPC6576-100GEA 203 EUR
APOSPC6576-100G APOSPC6576-25G APOSPC6576-5G
4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride 98%
4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃BrClF₃O₂S
MW: 323.52
Melting Pt: 54…58 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094392
CAS Number: 176225-10-8
UN: 3261
ADR: 8 III