HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Supplier: Apollo Scientific
Warning
Synonyms: HBTU, 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)
PC6587-100G PC6587-1KG PC6587-500G
APOSPC6587-100GEA 21.9 EUR
APOSPC6587-100G APOSPC6587-1KG APOSPC6587-500G
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Peptide coupling reagent.
Formula: C₁₁H₁₆F₆N₅OP
MW: 379.2 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00075445
CAS Number: 94790-37-1
EINECS: 423-020-5