3-Chloro-5-fluoro-4-hydroxybenzaldehyde
Supplier: Apollo Scientific
Danger
PC6621-1G PC6621-5G
APOSPC6621-1GEA 46.4 EUR
APOSPC6621-1G APOSPC6621-5G
3-Chloro-5-fluoro-4-hydroxybenzaldehyde
3-Chloro-5-fluoro-4-hydroxybenzaldehyde
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 134…138 °C
Storage Temperature: Ambient
CAS Number: 870704-13-5
UN: 2811
ADR: 6.1 III