5-Bromo-4-fluoro-2-methylaniline
Supplier: Apollo Scientific
Warning
PC6623-1G PC6623-25G PC6623-5G
APOSPC6623-1GEA 14.7 EUR
APOSPC6623-1G APOSPC6623-25G APOSPC6623-5G
5-Bromo-4-fluoro-2-methylaniline
5-Bromo-4-fluoro-2-methylaniline
Formula: C₇H₇BrFN
MW: 204.04 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD05865218
CAS Number: 627871-16-3