Tetrabutylammonium fluoride trihydrate 97%
Supplier: Apollo Scientific
Danger
PC6633T-100G PC6633T-500G PC6633T-1KG
APOSPC6633T-100GEA 21.9 EUR
APOSPC6633T-100G APOSPC6633T-500G APOSPC6633T-1KG
Tetrabutylammonium fluoride trihydrate 97%
Tetrabutylammonium fluoride trihydrate
Technical bulletin available
Formula: C₁₆H₃₆FN·3H₂O
Melting Pt: 54…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00149981
CAS Number: 87749-50-6
EINECS: 207-057-2
UN: 1759
ADR: 8,III