1,1,2,2-Tetrachlorotetrafluorocyclobutane 97%

Supplier: Apollo Scientific
Warning

PC6640-25G PC6640-5G
APOSPC6640-25GEA 503 EUR
APOSPC6640-25G APOSPC6640-5G
1,1,2,2-Tetrachlorotetrafluorocyclobutane 97%
1,1,2,2-Tetrachlorotetrafluorocyclobutane
Formula: C₄Cl₄F₄
Boiling Pt: 131…133 °C
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00039440
CAS Number: 336-50-5
EINECS: 206-409-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR