1-(3-Fluorophenyl)propanol
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-Fluorophenyl)propanol, alpha-Ethyl-3-fluorobenzyl alcohol
PC6641-1G PC6641-5G
APOSPC6641-1GEA 58 EUR
APOSPC6641-1G APOSPC6641-5G
1-(3-Fluorophenyl)propanol
1-(3-Fluorophenyl)propanol
Formula: C₉H₁₁FO
MW: 154.18 g/mol
Boiling Pt: 115 °C/17mmHg
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD08062380
CAS Number: 701-38-2