1,1,2,2-Tetrafluoropropane 97%
Supplier: Apollo Scientific
Danger
Synonyms: FC-254cb
PC6645-5G PC6645-10G
APOSPC6645-5GEA 590 EUR
APOSPC6645-5G APOSPC6645-10G
1,1,2,2-Tetrafluoropropane 97%
1,1,2,2-Tetrafluoropropane
Formula: C₃H₄F₄
MW: 116.06 g/mol
Boiling Pt: –0.8 °C
Melting Pt: –121 °C
Density: 1.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00798128
CAS Number: 40723-63-5
UN: 3161
ADR: 2.1