1-Bromo-2,6-bis(trifluoromethyl)benzene
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-1,3-bis(trifluoromethyl)benzene, 1,3-Bis(trifluoromethyl)-2-bromobenzene
PC6654-25G
APOSPC6654-25GEA 119 EUR
APOSPC6654-25G
1-Bromo-2,6-bis(trifluoromethyl)benzene
1-Bromo-2,6-bis(trifluoromethyl)benzene
Formula: C₈H₃BrF₆
MW: 293.0038 g/mol
Boiling Pt: 86 °C/29mmHg
Melting Pt: 51…52 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236210
CAS Number: 118527-30-3