1H,1H,9H,9H-Perfluoro-1,9-nonanediol 90%
Supplier: Apollo Scientific
Warning
Synonyms: 1H,1H,9H,9H-Perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluoro-1,9-nonandiol
PC6658-25G PC6658-5G
APOSPC6658-25GEA 306 EUR
APOSPC6658-25G APOSPC6658-5G
1H,1H,9H,9H-Perfluoro-1,9-nonanediol 90%
1H,1H,9H,9H-Tetradecafluoro-1,9-nonandiol
Formula: C₉H₆F₁₄O₂
MW: 412.12 g/mol
Boiling Pt: 140 °C/5mmHg
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00153275
CAS Number: 203303-01-9