4-(3-Fluorophenoxy)piperidine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 4-(3-Fluorophenoxy)piperidinehydrochloride
PC6669-1G PC6669-250MG
APOSPC6669-1GEA 105 EUR
APOSPC6669-1G APOSPC6669-250MG
4-(3-Fluorophenoxy)piperidine hydrochloride
4-(3-Fluorophenoxy)piperidine hydrochloride
Formula: C₁₁H₁₅ClFNO
MW: 231.690 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08445760
CAS Number: 3202-36-6