2,2,2,4'-Tetrafluoroacetophenone 99%
Supplier: Apollo Scientific
|
Danger
|
PC6681-100G
PC6681-25G
PC6681-5G
APOSPC6681-100GEA
214
EUR
APOSPC6681-100G
APOSPC6681-25G
APOSPC6681-5G
2,2,2,4'-Tetrafluoroacetophenone 99%
2,2,2,4'-Tetrafluoroacetophenone
Formula:
C₈H₄F₄O Boiling Pt: 66…67 °C/34mmHg Melting Pt: 24…27 °C Density: 1.37 g/cm³ (25 °C) Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042393 CAS Number: 655-32-3 EINECS: 211-509-4 UN: 1325 ADR: 4.1,III |