2,2,2,3'-Tetrafluoroacetophenone 99%
Supplier: Apollo Scientific
Danger
Synonyms: 2,2,2-Trifluoro-1-(3-fluorophenyl)ethanone
PC6681D-25G PC6681D-5G
APOSPC6681D-25GEA 556 EUR
APOSPC6681D-25G APOSPC6681D-5G
2,2,2,3'-Tetrafluoroacetophenone 99%
2,2,2,3'-Tetrafluoroacetophenone
Formula: C₈H₄F₄O
Boiling Pt: 79 °C/50mmHg
Density: 1.372 g/cm³ (25 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00039236
CAS Number: 708-64-5
EINECS: 211-903-6
UN: 1224
ADR: 3,II