4-Fluoro-2-(trifluoromethyl)phenol
Supplier: Apollo Scientific
Danger
Synonyms: 5-Fluoro-2-hydroxybenzotrifluoride
PC6683-5G PC6683-25G
APOSPC6683-5GEA 55.1 EUR
APOSPC6683-5G APOSPC6683-25G
4-Fluoro-2-(trifluoromethyl)phenol
4-Fluoro-2-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Melting Pt: 51…55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061290
CAS Number: 130047-19-7
UN: 3261
ADR: 8 III