2,3,4,5-Tetrafluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC6685-1G PC6685-5G PC6685-25G
APOSPC6685-1GEA 21.9 EUR
APOSPC6685-1G APOSPC6685-5G APOSPC6685-25G
2,3,4,5-Tetrafluoroaniline 98%
2,3,4,5-Tetrafluoroaniline
Formula: C₆H₃F₄N
MW: 165.09 g/mol
Boiling Pt: 78 °C/13mmHg
Melting Pt: 27…29 °C
Density: 1.53 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00025153
CAS Number: 5580-80-3