4'-Fluorobutyrophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Fluorophenyl)butan-1-one
PC6692-25G
APOSPC6692-25GEA 423 EUR
APOSPC6692-25G
4'-Fluorobutyrophenone 97%
4'-Fluorobutyrophenone
Formula: C₁₀H₁₁FO
MW: 166.2 g/mol
Boiling Pt: 230…232 °C
Melting Pt: 35…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00017967
CAS Number: 582-83-2
EINECS: 209-492-3