1,1-Dichloro-2,2,2-trifluoroethyldifluoromethyl ether 98%
Supplier: Apollo Scientific
Warning
PC6699-25G PC6699-5G
APOSPC6699-25GEA 584 EUR
APOSPC6699-25G APOSPC6699-5G
1,1-Dichloro-2,2,2-trifluoroethyldifluoromethyl ether 98%
1,1-Dichloro-2,2,2-trifluoroethyldifluoromethyl ether
Formula: C₃HCl₂F₅O
MW: 218.94 g/mol
Boiling Pt: 60…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01320775
CAS Number: 32778-07-7
UN: 1993
ADR: 3,III