(2-Chloro-4-fluorophenyl)acetamide 97%
Supplier: Apollo Scientific
|
Warning
|
PC6711-1G
PC6711-250MG
APOSPC6711-1GEA
122
EUR
APOSPC6711-1G
APOSPC6711-250MG
(2-Chloro-4-fluorophenyl)acetamide 97%
(2-Chloro-4-fluorophenyl)acetamide
Formula:
C₈H₇ClFNO Melting Pt: 171…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD02180958 CAS Number: 306937-35-9 |