1,2,3,5-Tetrafluorobenzene 97+%
Supplier: Apollo Scientific
Danger
PC6720-5G PC6720-1G PC6720-25G
APOSPC6720-5GEA 102 EUR
APOSPC6720-5G APOSPC6720-1G APOSPC6720-25G
1,2,3,5-Tetrafluorobenzene 97+%
1,2,3,5-Tetrafluorobenzene
Formula: C₆H₂F₄
MW: 150.08 g/mol
Boiling Pt: 82…83 °C
Melting Pt: –48 °C
Density: 1.393 g/cm³ (25 °C)
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000302
CAS Number: 2367-82-0
EINECS: 219-126-4
UN: 1993
ADR: 3,II