1,2,4,5-Tetrafluorobenzene 99%
Supplier: Apollo Scientific
Danger
PC6730-10G PC6730-250G PC6730-50G
APOSPC6730-10GEA 21.9 EUR
APOSPC6730-10G APOSPC6730-250G APOSPC6730-50G
1,2,4,5-Tetrafluorobenzene 99%
1,2,4,5-Tetrafluorobenzene
Formula: C₆H₂F₄
MW: 150.08 g/mol
Boiling Pt: 90 °C
Melting Pt: 44319 °C
Density: 1.344 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00000307
CAS Number: 327-54-8
EINECS: 206-319-3
UN: 1993
ADR: 3,II