2,3,4,5-Tetrafluorobenzoic acid 99%
Supplier: Apollo Scientific
Warning
PC6735-25G PC6735-500G PC6735-100G PC6735-1KG
APOSPC6735-25GEA 0 EUR
APOSPC6735-25G APOSPC6735-500G APOSPC6735-100G APOSPC6735-1KG
2,3,4,5-Tetrafluorobenzoic acid 99%
2,3,4,5-Tetrafluorobenzoic acid
Formula: C₇H₂F₄O₂
MW: 194.09 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00009613
CAS Number: 1201-31-6