(S)-(+)-3-Fluoropyrrolidine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: (S)-3-Fluoropyrrolidine hydrochloride
PC6738-1G PC6738-500MG PC6738-5G
APOSPC6738-1GEA 58 EUR
APOSPC6738-1G APOSPC6738-500MG APOSPC6738-5G
(S)-(+)-3-Fluoropyrrolidine hydrochloride
(S)-(+)-3-Fluoropyrrolidine hydrochloride
Formula: C₄H₉ClFN
MW: 125.57 g/mol
Melting Pt: 183…187 °C
Storage Temperature: Ambient
MDL Number: MFCD04038718
CAS Number: 136725-53-6