3-Bromo-5-fluoropyridine 97%
Supplier: Apollo Scientific
Danger
PC6747-100G PC6747-1G PC6747-50G
APOSPC6747-100GEA 277 EUR
APOSPC6747-100G APOSPC6747-1G APOSPC6747-50G
3-Bromo-5-fluoropyridine 97%
3-Bromo-5-fluoropyridine
Formula: C₅H₃BrFN
Boiling Pt: 78 °C/11mmHg
Melting Pt: 24…28 °C
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD04112555
CAS Number: 407-20-5