(1,1,2,2-Tetrafluoroethoxy)benzene 98%
Supplier: Apollo Scientific
Warning
PC6767-100G PC6767-25G PC6767-5G
APOSPC6767-100GEA 609 EUR
APOSPC6767-100G APOSPC6767-25G APOSPC6767-5G
(1,1,2,2-Tetrafluoroethoxy)benzene 98%
(1,1,2,2-Tetrafluoroethoxy)benzene
Formula: HF₂CCF₂OC₆H₅
MW: 194.13 g/mol
Boiling Pt: 152…153 °C
Density: 1.275 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00042102
CAS Number: 350-57-2
EINECS: 206-505-4
UN: 1993
ADR: 3 III