3-Fluoro-5-nitrobenzonitrile
Supplier: Apollo Scientific
Danger
PC6771-1G PC6771-5G
APOSPC6771-1GEA 223 EUR
APOSPC6771-1G APOSPC6771-5G
3-Fluoro-5-nitrobenzonitrile
3-Fluoro-5-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
MW: 166.11 g/mol
Melting Pt: 49…54 °C
Density: 1.41 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD05864356
CAS Number: 110882-60-5
UN: 3439
ADR: 6.1 III