1-Nitro-2,3,4,5-tetrafluorobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3,4,5-Tetrafluoronitrobenzene, 1,2,3,4-Tetrafluoro-5-nitrobenzene
PC6809-5G PC6809-1G PC6809-25G
APOSPC6809-5GEA 51 EUR
APOSPC6809-5G APOSPC6809-1G APOSPC6809-25G
1-Nitro-2,3,4,5-tetrafluorobenzene 99%
1-Nitro-2,3,4,5-tetrafluorobenzene
Formula: C₆HF₄NO₂
MW: 195.07 g/mol
Boiling Pt: 80 °C/24mmHg
Density: 1.618 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00014686
CAS Number: 5580-79-0
EINECS: 226-972-8