1,2,3,5-Tetrafluoro-4-nitrobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3,4,6-Tetrafluoronitrobenzene
PC6810-1G PC6810-5G PC6810-10G
APOSPC6810-1GEA 120 EUR
APOSPC6810-1G APOSPC6810-5G APOSPC6810-10G
1,2,3,5-Tetrafluoro-4-nitrobenzene 97%
1,2,3,5-Tetrafluoro-4-nitrobenzene
Formula: C₆HF₄NO₂
MW: 195.07 g/mol
Boiling Pt: 78…79 °C/20mmHg
Melting Pt: –5 °C
Density: 1.511 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00011687
CAS Number: 314-41-0
EINECS: 206-246-7