4'-Hydroxy-3'-(trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Warning
PC6845-1G PC6845-250MG
APOSPC6845-1GEA 63.8 EUR
APOSPC6845-1G APOSPC6845-250MG
4'-Hydroxy-3'-(trifluoromethyl)acetophenone
4'-Hydroxy-3'-(trifluoromethyl)acetophenone
Formula: C₉H₇F₃O₂
Melting Pt: 174…179 °C
Storage Temperature: Ambient
MDL Number: MFCD01091005
CAS Number: 149105-11-3