3-Amino-2,5,6-trifluorobenzoic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC6848-1G
PC6848-5G
PC6848-25G
APOSPC6848-1GEA
79.9
EUR
APOSPC6848-1G
APOSPC6848-5G
APOSPC6848-25G
3-Amino-2,5,6-trifluorobenzoic acid 98%
Formula:
C₇H₄F₃NO₂ Melting Pt: 175…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075469 CAS Number: 133622-65-8 |