4-Chloro-3-fluorophenylboronic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-3-fluorobenzeneboronic acid, (4-Chloro-3-fluorophenyl)metaboric acid, (4-Chloro-3-fluorophenyl)boronic acid, Dihydroxy(4-chloro-3-fluorophenyl)borane
PC6912-1G PC6912-25G PC6912-5G
APOSPC6912-1GEA 21.9 EUR
APOSPC6912-1G APOSPC6912-25G APOSPC6912-5G
4-Chloro-3-fluorophenylboronic acid 98%
4-Chloro-3-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
Melting Pt: 198…204 °C
Storage Temperature: Ambient
MDL Number: MFCD01319010
CAS Number: 137504-86-0