2-Bromo-1-(4-bromo-2-fluorophenyl)ethanone 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Bromo-2-fluorophenacyl bromide
PC6936-1G PC6936-5G PC6936-10G
APOSPC6936-1GEA 174 EUR
APOSPC6936-1G APOSPC6936-5G APOSPC6936-10G
2-Bromo-1-(4-bromo-2-fluorophenyl)ethanone 98%
2-Bromo-1-(4-bromo-2-fluorophenyl)ethanone
Formula: C₈H₅Br₂FO
MW: 295.93 g/mol
Melting Pt: 79…80 °C
Storage Temperature: Ambient
MDL Number: MFCD11847024
CAS Number: 869569-77-7
UN: 1759
ADR: 8,III