Thallium(I) fluoride 99%
Supplier: Apollo Scientific
Danger
PC6942-25G PC6942-100G
APOSPC6942-25GEA 357 EUR
APOSPC6942-25G APOSPC6942-100G
Thallium(I) fluoride 99%
Thallium (I) fluoride
Formula: FTl
MW: 223.38 g/mol
Boiling Pt: 655 °C
Melting Pt: 327 °C
Density: 8.23 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049607
CAS Number: 7789-27-7
UN: 1707
ADR: 6.1,II