1-(2-Thenoyl)-3,3,3-trifluoroacetone
Supplier: Apollo Scientific
Warning
Synonyms: (TTA), Thenoyltrifluoroacetone, 4,4,4-Trifluoro-1-(2-thienyl)butane-1,3-dione, TTFA, 2-Thenoyltrifluoroacetone, (2-Thenoyl)-ω,ω,ω-trifluoroacetone
PC6950-100G PC6950-250G PC6950-25G PC6950-BULK
APOSPC6950-100GEA 53.8 EUR
APOSPC6950-100G APOSPC6950-250G APOSPC6950-25G APOSPC6950-BULK
1-(2-Thenoyl)-3,3,3-trifluoroacetone
1-(2-Thenoyl)-3,3,3-trifluoroacetone
Formula: C₈H₅F₃O₂S
Boiling Pt: 96…98 °C/8mmHg
Melting Pt: 42…43 °C
Flash Pt: 111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005445
CAS Number: 326-91-0
EINECS: 206-316-7