2,5-Dichloro-3-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
PC6960-1G PC6960-500MG PC6960-5G
APOSPC6960-1GEA 219 EUR
APOSPC6960-1G APOSPC6960-500MG APOSPC6960-5G
2,5-Dichloro-3-(trifluoromethyl)pyridine
2,5-Dichloro-3-(trifluoromethyl)pyridine
Formula: C₆H₂Cl₂F₃N
MW: 215.9880 g/mol
Boiling Pt: 80…82 °C/26mmHg
Storage Temperature: Ambient
MDL Number: MFCD00042244
CAS Number: 70158-59-7