3,4,5-Trichloro-α,α,α-trifluorotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4,5-Trichlorobenzotrifluoride, 1,2,3-Trichloro-5-(trifluoromethyl)benzene
PC7009-100G PC7009-25G PC7009-500G
APOSPC7009-100GEA 31.9 EUR
APOSPC7009-100G APOSPC7009-25G APOSPC7009-500G
3,4,5-Trichloro-α,α,α-trifluorotoluene 98%
3,4,5-Trichloro-α,α,α-trifluorotoluene
Formula: C₇H₂Cl₃F₃
MW: 249.45 g/mol
Boiling Pt: 200…206 °C
Melting Pt: –10…–8 °C
Density: 1.6 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00035985
CAS Number: 50594-82-6
EINECS: 256-636-6