2-Bromo-5-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC7010-25G PC7010-5G PC7010-100G
APOSPC7010-25GEA 94.4 EUR
APOSPC7010-25G APOSPC7010-5G APOSPC7010-100G
2-Bromo-5-(trifluoromethyl)benzaldehyde 97%
2-Bromo-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄BrF₃O
MW: 253.02 g/mol
Boiling Pt: 50…55 °C
Storage Temperature: Ambient
MDL Number: MFCD04973760
CAS Number: 102684-91-3