2-Fluoro-3-formylpyridine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoronicotinaldehyde
PC7036-25G PC7036-5G
APOSPC7036-25GEA 165 EUR
APOSPC7036-25G APOSPC7036-5G
2-Fluoro-3-formylpyridine 95%
2-Fluoro-3-formylpyridine
Formula: C₆H₄FNO
MW: 125.1 g/mol
Boiling Pt: 50 °C
Density: 1.25 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD03095270
CAS Number: 36404-90-7